EMAIL THIS PAGE TO A FRIEND
H7881 Sigma-Aldrich

Aluminum oxide

5 μm mean particle size

Synonym: Alumina

Purchase

Properties

Related Categories Aluminum, Materials Science, Metal and Ceramic Science, Oxides
InChI Key   TWNQGVIAIRXVLR-UHFFFAOYSA-N
mean particle size   5 μm
surface area   70 m2/g
mp   2040 °C(lit.)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD1200000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?