EMAIL THIS PAGE TO A FRIEND
I1015 Sigma-Aldrich

Immersion oil

viscosity 21,000 cSt (lit.)

Purchase

Properties

Related Categories Cell Biology, G-H-I, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
viscosity   21,000 cSt(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
325.4 °F
Flash Point(C) 
163 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?