• USA Home
  • I8405 - Insulin from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
I8405 Sigma-Aldrich

Insulin from bovine pancreas

meets USP testing specifications

Purchase

Properties

Related Categories Analytical/Chromatography, Biochemicals and Reagents, Insulin and related, Pharmacopoeia, Pharmacopoeia A-Z,
form   solid
suitability   meets USP testing specifications
storage temp.   −20°C
Gene Information   cow ... INS(280829)

Description

Biochem/physiol Actions

Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Quality

Purified

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?