• USA Home
  • IRMM502B - Nuclear reference material 237NpO2 B

EMAIL THIS PAGE TO A FRIEND
IRMM502B Sigma-Aldrich

Nuclear reference material 237NpO2 B

IRMM® certified Reference Material

Synonym: Neptunium(IV) oxide nuclear reference material, Neptunium(IV) oxide

  • CAS Number 58670-16-9

  • Empirical Formula (Hill Notation) NpO2

  • Molecular Weight 269.00

  •  MDL number MFCD04974579

Purchase

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

IRMM is a registered trademark of European Commission

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?