• USA Home
  • IRMMIFCC456 - α-Amylase (catalytic concentration)

EMAIL THIS PAGE TO A FRIEND
IRMMIFCC456 Sigma-Aldrich

α-Amylase (catalytic concentration)

IRMM® certified Reference Material

Synonym: α-Amylase from Bacillus amyloliquefaciens

Purchase

Description

Analysis Note

For more information please see:
IRMMIFCC456

Legal Information

IRMM is a registered trademark of European Commission

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?