• USA Home
  • LRWM - LR white embedding media Medium

EMAIL THIS PAGE TO A FRIEND
LRWM Sigma-Aldrich

LR white embedding media Medium

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, J-K-L, Stains & Dyes, A to Z, Stains and Dyes More...
storage temp.   2-8°C

Description

Other Notes

This product has been replaced by 62662-SIGMA | LR White embedding kit

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications

Standard component

Description

Product #

Add to Cart

LR White accelerator 2 mL SDS    
LR White resin medium 100 g SDS L9774
stabilized benzoyl peroxide 1 mL/vial SDS B5907
Safety & Documentation

Safety Information

RIDADR 
UN 3106 5.2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?