CAS Number 13446-34-9
Linear Formula MnCl2 · 4H2O
Molecular Weight 197.91
EC Number 231-869-6
MDL number MFCD00149792
PubChem Substance ID 24896860
Popular Documents: Product Information Sheet (PDF)
| grade | ReagentPlus® |
| assay | ≥99% |
| mp | 58 °C(lit.) |
100, 500 g in poly bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?