• USA Home
  • M6380 - Morphine-d3 hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
M6380 Sigma-Aldrich

Morphine-d3 hydrochloride solution

100 μg/mL in methanol, analytical standard, for drug analysis

Purchase

Properties

Related Categories Additional Standards, Alkaloids, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard, for drug analysis
InChI Key   XCKKIKBIPZJUET-MXXLTCJRSA-N
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   100 μg/mL in methanol
  100 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?