CAS Number 7791-18-6
Linear Formula MgCl2 · 6H2O
Molecular Weight 203.30
EC Number 232-094-6
MDL number MFCD00149781
PubChem Substance ID 24897325
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | 99.0-102.0% |
| impurities | ≤0.005% Insoluble matter |
| anion traces | nitrate (NO3-): ≤0.001% |
| phosphate (PO43-): ≤5 ppm | |
| sulfate (SO42-): ≤0.002% | |
| cation traces | Ba: ≤0.005% |
1 kg in poly bottle
100, 500 g in poly bottle
2.5, 12 kg in poly drum
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?