• USA Home
  • MURSTISSA1 - Antarctic sediment (trace elements)

EMAIL THIS PAGE TO A FRIEND
MURSTISSA1 Sigma-Aldrich

Antarctic sediment (trace elements)

reference material certified by ISS (Istituto Superiore di Sanita, Rome)

Purchase

Properties

grade   certified reference material
  reference material certified by ISS (Istituto Superiore di Sanita, Rome)
form   neat
application(s)   ICP/AAS: suitable
format   neat

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

BCR320R Channel sediment (trace elements), BCR® certified Reference Material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?