• USA Home
  • N6376 - Nitro Blue Diformazan

EMAIL THIS PAGE TO A FRIEND
N6376 Sigma-Aldrich

Nitro Blue Diformazan

Synonym: 1,1′-(3,3′-Dimethoxy-4,4′-biphenylylene)bis[5-(p-nitrophenyl)-3-phenyl-formazan], 5,5′-Di-p-nitrophenyl-3,3′-diphenyl-1,1′-(3,3′-dimethoxy-4,4′-diphenylene) diformazan, Diformazan of nitro blue tetrazolium

  • CAS Number 16325-01-2

  • Empirical Formula (Hill Notation) C40H32N10O6

  • Molecular Weight 748.75

  •  MDL number MFCD00047719

Purchase

Properties

InChI Key   NHKRPSVRVUKAJC-LTWSLYJCSA-N
mp   200-270 °C

Description

Quality

Very low monoformazan impurity.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?