EMAIL THIS PAGE TO A FRIEND
N8153 Sigma-Aldrich

3-Nitrosobenzamide

analytical standard, for drug analysis

Purchase

Properties

Related Categories Additional Drugs, Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Structure,
grade   analytical standard, for drug analysis
InChI Key   OZUBORKYZRYLSQ-UHFFFAOYSA-N
assay   ≥98%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1759 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?