• USA Home
  • NIST1710 - Aluminum alloy 3004
EMAIL THIS PAGE TO A FRIEND
NIST1710 Sigma-Aldrich
Aluminum alloy 3004
NIST SRM 1710
Purchase
Description
General description
This Standard Reference Material (SRM) is in the form of a disk approximately 63 mm (2 l/2 in) in diameter and 19mm (3/4 in) thick, and is intended primarily for use with optical emission and x-ray fluorescence methods of analysis. For more information, please refer to the SDS and COA.
SRM 1710_cert SRM 1710 _SDS
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?