• USA Home
  • NIST2113 - Dynamic impact force verification specimens
EMAIL THIS PAGE TO A FRIEND
NIST2113 Sigma-Aldrich
Dynamic impact force verification specimens
NIST SRM 2113, nominal 33 kN
Purchase
Description
General description
This Standard Reference Material (SRM) is intended primarily for the verification of the maximum force measured dynamically using an instrumented Charpy impact machine in accordance with the current ASTM Standard E2298 or ISO Standard 14556 . A unit of SRM 2113 consists of a set of four specimens needed to perform a single verification. SRM 2113 can also be used to verify the absorbed energy scale of the impact machine at the low energy level. for more information, please refer to the SDS and COA.
SRM 2113_cert SRM 2113 _SDS
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?