EMAIL THIS PAGE TO A FRIEND
O1625 Sigma-Aldrich

Orange G

Dye content ≥60 %

Synonym: 1-Phenylazo-2-naphthol-6,8-disulfonic acid disodium salt, 7-Hydroxy-8-phenylazo-1,3-naphthalenedisulfonic acid disodium salt, Acid Orange 10, Wool Orange 2G

  • CAS Number 1936-15-8

  • Empirical Formula (Hill Notation) C16H10N2Na2O7S2

  • Molecular Weight 452.37

  •  Beilstein Registry Number 4120705

  •  Colour Index Number 16230

  •  EC Number 217-705-6

  •  MDL number MFCD00012457

  •  PubChem Substance ID 329818898

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, O, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   HSXUHWZMNJHFRV-QIKYXUGXSA-L
composition   Dye content, ≥60%

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QJ6500000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?