EMAIL THIS PAGE TO A FRIEND
P0343 Sigma-Aldrich

TWEEN® 80 solution

10%, low peroxide

Synonym: Polyethylene glycol sorbitan monooleate solution, Polyoxyethylenesorbitan Monooleate

Purchase

Description

Other Notes

Contains BHT as an antioxidant

Legal Information

TWEEN is a registered trademark of Croda International PLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WG2934000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?