EMAIL THIS PAGE TO A FRIEND
P1338 Sigma-Aldrich

Potassium chlorate

BioXtra, ≥99.0%

Purchase

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Potassium, Synthetic Reagents
InChI Key   VKJKEPKFPUWCAS-UHFFFAOYSA-M
product line   BioXtra
assay   ≥99.0%
impurities   <0.0005% Phosphorus (P)
  <0.005% Insoluble matter
mp   356 °C(lit.)
solubility   H2O: soluble1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): <0.001%
  sulfate (SO42-): <0.002%
cation traces   Al: <0.0005%
  Ca: <0.0005%
  Cu: <0.0005%
  Fe: <0.0005%
  Mg: <0.0005%
  NH4+: <0.05%
  Na: <0.005%
  Pb: <0.0005%
  Zn: <0.0005%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1485 5.1 / PGII
WGK Germany 
2
RTECS 
FO0350000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?