EMAIL THIS PAGE TO A FRIEND
P2565 Sigma-Aldrich

TWEEN® 21

Synonym: Polyoxyethylenesorbitan monolaurate

Purchase

Properties

Related Categories Biochemicals and Reagents, Core Bioreagents, Detergents, Detergents N to Z, Non-Ionic Detergents,
description   non-ionic
composition   Lauric acid, ≥40% (balance primarily myristic and palmitic acids)

Description

Legal Information

TWEEN is a registered trademark of Croda International PLC

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TR7400000
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?