P3289 Sigma-Aldrich

Potassium hexacyanoferrate(II) trihydrate

ACS reagent, 98.5-102.0%

DOWNLOAD MSDS (PDF)

Synonym: Potassium ferrocyanide, Yellow prussiate

  • CAS Number 14459-95-1

  • Linear Formula K4Fe(CN)6 · 3H2O

  • Molecular Weight 422.39

  •  EC Number 237-722-2

  •  MDL number MFCD00167023

  •  PubChem Substance ID 24897881

Properties

Related Categories ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Core Bioreagents, Essential Chemicals,
grade   ACS reagent
assay   98.5-102.0%
impurities   ≤0.005% Insoluble matter
mp   70 °C(lit.)
anion traces   chloride (Cl-): ≤0.01%
  sulfate (SO42-): passes test (limit about 0.01%)

Description

Packaging

5, 100, 500 g in poly bottle

Price and Availability

Transfection Reagents


TransfectionFind all the necessary reagents to perfom
your Transfection research in one place.






Customers Also Viewed

Potassium ferricyanide(III)

powder, <10 μm, 99%

Potassium hexacyanoferrate(II) trihydrate

≥99.99% trace metals basis

Potassium hexacyanoferrate(II) trihydrate

ReagentPlus®, ≥98.5%

Potassium hexacyanoferrate(II) trihydrate

BioUltra, ≥99.5% (RT)

Potassium hexacyanoferrate(III)

ACS reagent, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?