EMAIL THIS PAGE TO A FRIEND
P3691 Sigma-Aldrich

Potassium bromide

Purchase

Properties

Related Categories Essential Chemicals, Potassium Salts, Research Essentials, Salts
vapor pressure   <0.01 mmHg ( 20 °C)
  1 mmHg ( 795 °C)
InChI Key   IOLCXVTUBQKXJR-UHFFFAOYSA-M
mp   734 °C(lit.)

Description

Application

May be used for removal of peripheral membrane proteins.

Quality

Photographic Grade

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TS7650000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?