CAS Number 7447-40-7
Linear Formula KCl
Molecular Weight 74.55
EC Number 231-211-8
MDL number MFCD00011360
PubChem Substance ID 24898470
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Buffers, Biological Buffers, Buffers A to Z, Chemical Synthesis, |
| grade | ACS reagent |
| assay | 99.0-100.5% |
| impurities | ≤0.005% Insoluble matter |
| pH | 5.4-8.6 (25 °C, 5% in solution) |
| mp | 770 °C(lit.) |
| density | 1.98 g/mL at 25 °C(lit.) |
| anion traces | bromide (Br-): ≤0.01% |
May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.
1, 2.5 kg in poly bottle
25, 500 g in poly bottle
5, 12 kg in poly drum
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?