• USA Home
  • P4838 - Peptone Primatone® HS

EMAIL THIS PAGE TO A FRIEND
P4838 Sigma-Aldrich

Peptone Primatone® HS

for microbiology

Synonym: Peptone from animal tissue

Purchase

Description

Legal Information

Primatone is a registered trademark of Kerry Group

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P8388 Primatone®, Meat protein enzymatic hydrolysate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?