CAS Number 584-08-7
Linear Formula K2CO3
Molecular Weight 138.21
Beilstein Registry Number 4267587
EC Number 209-529-3
MDL number MFCD00011382
PubChem Substance ID 24898711
Popular Documents: Specification Sheet (PDF)
| Related Categories | Adsorbents, Filter Aids and Drying Agents, Buffers / Buffer Salts, Buffers / Buffer salts, Chemical Synthesis, Core Bioreagents, |
| product line | BioXtra |
| assay | ≥99.0% |
| impurities | ≤0.0005% Phosphorus (P) |
| ≤0.1% Insoluble matter | |
| mp | 891 °C(lit.) |
| solubility | H2O: soluble1 M, clear, colorless |
| anion traces | chloride (Cl-): ≤0.005% |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?