• USA Home
  • P6949 - Potassium antimony(III) tartrate hydrate

EMAIL THIS PAGE TO A FRIEND
P6949 Sigma-Aldrich

Potassium antimony(III) tartrate hydrate

99% (titration)

Synonym: Antimony potassium tartrate

  • CAS Number 331753-56-1

  • Empirical Formula (Hill Notation) C8H4K2O12Sb2 · xH2O

  • Molecular Weight 613.83 (anhydrous basis)

  •  MDL number MFCD00149437

  •  PubChem Substance ID 24898807

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acid Salts, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   BBUFTTIHMIIQLI-UHFFFAOYSA-J
assay   99% (titration)
optical activity   [α]20/D +141°, c = 2 in H2O
density   2.6 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by 244791-ALDRICH | Potassium antimony(III) tartrate hydrate ≥99%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 1551 6.1 / PGIII
WGK Germany 
3
RTECS 
CC6825000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

244791 Potassium antimony(III) tartrate hydrate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?