EMAIL THIS PAGE TO A FRIEND
P7143 Sigma-Aldrich

Picrotin

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alkaloids, Analytical Standards, Analytical/Chromatography, Chemical Structure,
grade   analytical standard
InChI Key   RYEFFICCPKWYML-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Non-toxic component of picrotoxin

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TJ8800000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?