P7754 Sigma-Aldrich

Acetylacetone

ReagentPlus®, ≥99%

DOWNLOAD MSDS (PDF)

Synonym: 2,4-Pentanedione

Properties

Related Categories A-AM, Bioactive Small Molecules, Cell Biology, Essential Chemicals, Inorganic Salts, Solutions and Reagents,
grade   ReagentPlus®
vapor density   3.5 (vs air)
vapor pressure   6 mmHg ( 20 °C)
assay   ≥99%
autoignition temp.   662 °F
expl. lim.   11.4 %
refractive index   n20/D 1.452(lit.)
bp   140.4 °C(lit.)
mp   −23 °C(lit.)
density   0.975 g/mL at 25 °C(lit.)
Gene Information   human ... ACHE(43), BCHE(590), CES1(1066)

Description

Application

Transition-metal chelating agent

Packaging

1 L in glass bottle

100, 500 mL in glass bottle

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

5α-Pregnane-3,20-dione (allo)

progesterone metabolite

Acetylacetone

analytical standard

Acetylacetone

puriss. p.a., ≥99.5% (GC)

Acetylacetone

produced by Wacker, ≥99.5% (GC)

Acetylacetone

≥99.3%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?