• USA Home
  • P8096 - Pancreatin from porcine pancreas

EMAIL THIS PAGE TO A FRIEND
P8096 Sigma-Aldrich

Pancreatin from porcine pancreas

meets USP testing specifications

Synonym: Pancreatin from hog pancreas

Purchase

Properties

Related Categories Analytical/Chromatography, Cell Culture, Pharmacopoeia, Pharmacopoeia A-Z, Pharmacopoeial Vitamins and Supplements,
activity   1 × USP specifications
suitability   meets USP testing specifications
storage temp.   −20°C

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Biochem/physiol Actions

Pancreatin contains enzymatic components including trypsin, amylase and lipase, ribonuclease, and protease, produced by the exocrine cells of the porcine pancreas. This combination of enzymes allows it to hydrolyze proteins, starch and fats. Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40°C, will digest not less than 25 times its weight of casein in 60 minutes at pH 7.5 at 40°C and will release not less than microequivalents of acid per min per mg pancreatin from olive oil at pH 9.0 at 37°C.

Price and Availability


Filter out uncertainty.

All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?