EMAIL THIS PAGE TO A FRIEND
P8471 Sigma-Aldrich

Pectin from apple

meets USP testing specifications

Synonym: Poly-D-galacturonic acid methyl ester

Purchase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Prebiotics

Prebiotic food components typically consist of polysaccharides and oligosaccharides. They can be found in milk (galactooligosaccharides or GOS) and plants with dietary fibers, and they are often prod...
BioFiles Edition 6.2
Keywords: Fermentation, Prebiotics

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

76282 Pectin from apple

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?