EMAIL THIS PAGE TO A FRIEND
Q4875 Sigma-Aldrich

Quin 2-AM

≥90%

Synonym: 2-[(2-Amino-5-methylphenoxy)methyl]-6-methoxy-8-aminoquinoline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl ester), 2-{[2-Bis(carboxymethyl)amino-5-methylphenoxy]-methyl}-6-methoxy-8-bis(carboxymethyl)aminoquinoline tetrakis(acetoxymethyl) ester

  • CAS Number 83104-85-2

  • Empirical Formula (Hill Notation) C38H43N3O18

  • Molecular Weight 829.76

  •  Beilstein Registry Number 8181991

  •  MDL number MFCD00010285

  •  PubChem Substance ID 24899332

Purchase

Description

Application

Cell permeable fluorescent probe for Ca2+ that gives a large shift in the UV absorption spectrum and a 20-fold increase in fluorescence upon Ca2+ binding

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Sensor Dyes

In 1998 R.F. Furchgott, L.J. Ignarro, and F. Murad received the Nobel Prize in Physiology and Medicine for their discoveries concerning nitric oxide as a signaling molecule in the cardiovascular syst...
Monika Baeumle
BioFiles 2011, 6.3, 23.
Keywords: Applications, Biochemistry, Biological processes, Cancer, Cardiovascular, Central Nervous System, Diseases, Extinction coefficient, Indicators, Inflammation, Methods, Microscopy, Neurotransmission, Sol-gel, Type

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?