EMAIL THIS PAGE TO A FRIEND
QC1085 Sigma-Aldrich

pH - Constant Value

certified reference material

Purchase

Properties

grade   certified reference material
packaging   pkg of 1 standard
mfr. no.   RTC, QCI-041
concentration   100% in H2O
format   matrix material

Description

General description

A single standard for direct measurement of pH.

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?