• USA Home
  • QC1312 - Alcohols in water - WS

EMAIL THIS PAGE TO A FRIEND
QC1312 Sigma-Aldrich

Alcohols in water - WS

certified reference material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Drinking Water Matrix CRMs,
grade   certified reference material
packaging   ampule of 1.2 mL
mfr. no.   RTC, QCO-230
concentration   100% in H2O
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   matrix material

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

tert-Butanol SDS    
Ethanol SDS    
2-Propanol SDS    
Methanol SDS    
4-Methyl-2-pentanone SDS    
Tetrahydrofuran SDS    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

PE1312 Alcohols in Water - WS, Proficiency Testing Material
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?