EMAIL THIS PAGE TO A FRIEND
QC1579 Sigma-Aldrich

PCBs - WS

certified reference material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Environmental Matrix CRMs (RTC),
grade   certified reference material
packaging   ampule of 2 mL
mfr. no.   RTC, QCO-003
concentration   100% in acetone
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   matrix material

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Aroclor 1260 SDS    
Decachlorobiphenyl SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1090 - class 3 - PG 2 - Acetone, solution
WGK Germany 
1
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

PE1579 PCB′s - WS, Proficiency Testing Material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?