EMAIL THIS PAGE TO A FRIEND
R0385 Sigma-Aldrich

Reactive Brown 10

practical grade

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Q-R, Stains & Dyes, A to Z, Stains and Dyes More...
grade   practical grade

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?