EMAIL THIS PAGE TO A FRIEND
R2303 Sigma-Aldrich

Rutin hydrate

95%

Synonym: Quercetin-3-rutinoside hydrate, Vitamin P hydrate

  • CAS Number 207671-50-9

  • Empirical Formula (Hill Notation) C27H30O16 · xH2O

  • Molecular Weight 610.52 (anhydrous basis)

  •  Beilstein Registry Number 75455

  •  EC Number 205-814-1

  •  MDL number MFCD01319140

Purchase

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

R5143 Rutin hydrate, ≥94% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?