EMAIL THIS PAGE TO A FRIEND
R8026 Sigma-Aldrich

RH 421

≥95%, solid

Synonym: 4-{4-[4-(Dipentylamino)phenyl]-1,3-butadienyl}-1-(4-sulfobutyl)pyridinium hydroxide

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Q-R, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   URNCTMHGJQSLOC-UHFFFAOYSA-N
assay   ≥95%
form   solid
color   red
solubility   DMF: soluble
  DMSO: soluble
  ethanol: soluble
storage temp.   2-8°C

Description

Biochem/physiol Actions

Chromogenic substrate for β-galactosidase that generates red or magenta precipitate on hydrolysis.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?