• USA Home
  • R9006 - Rhodamine B isothiocyanate–Dextran

EMAIL THIS PAGE TO A FRIEND
R9006 Sigma-Aldrich

Rhodamine B isothiocyanate–Dextran

average mol wt ~20,000

Synonym: RITC-Dextran

Purchase

Properties

mol wt   average mol wt ~20,000
extent of labeling   0.002-0.01 mol RITC per mol glucose
storage temp.   2-8°C

Description

Application

Dextran labeled with rhodamine B isothiocyanate for possible use in perfusion studies in animals.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?