S1179 Sigma-Aldrich

Silver nitrate

meets USP testing specifications

DOWNLOAD MSDS (PDF)

Synonym: Nitric acid silver(I) salt

  • CAS Number 7761-88-8

  • Linear Formula AgNO3

  • Molecular Weight 169.87

  •  EC Number 231-853-9

  •  MDL number MFCD00003414

Properties

Related Categories Pharmacopoeia, Pharmacopoeia A-Z, Pharmacopoeial Inorganics More...
vapor density   5.8 (vs air)
form   solid
mp   212 °C (dec.)(lit.)
suitability   meets USP testing specifications

Price and Availability

Customers Also Viewed

Silver nitrate

BioXtra, >99% (titration)

Silver nitrate

BioReagent, suitable for plant cell culture, >99% (titration)

Silver nitrate

99.9999% trace metals basis

Silver nitrate

ACS reagent, ≥99.0%

Silver nitrate

meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?