Synonym: Sodium periodate
CAS Number 7790-28-5
Linear Formula NaIO4
Molecular Weight 213.89
EC Number 232-197-6
MDL number MFCD00003534
PubChem Substance ID 24899499
Popular Documents: Specification Sheet (PDF)
| Related Categories | Chemical Synthesis, Core Bioreagents, Detection of Biotin Labeled Glycoproteins on Western Blots, Glycobiology, Glycoprotein Detection, |
| Quality Level | PREMIUM |
| assay | ≥99.0% |
| solubility | H2O: soluble50 mg/mL |
Live Chat and Frequently Asked Questions
are available for this Product.
Useful reagent for the oxidation of carbohydrates prior to labeling with biotin hydrazide
25, 100, 500 g in glass bottle
Customers Also Viewed
Analysis of proteins and glycoproteins on blots. Bayer, E.A.et. al. Meth. Enzymol. 184, 415, (1990)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?