Synonym: Calcined soda, Carbonic acid disodium salt, Soda ash
CAS Number 497-19-8
Linear Formula Na2CO3
Molecular Weight 105.99
Beilstein Registry Number 4154566
EC Number 207-838-8
MDL number MFCD00003494
PubChem Substance ID 24899511
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Biological Buffers, Biomolecular Mass Spectrometry, Buffers A to Z, Metabolic Labeling, Stable Isotopes More... |
| grade | ReagentPlus® |
| assay | ≥99.5% |
| pKa (25 °C) | (1) 6.37, (2) 10.25 (carbonic acid) |
| mp | 851 °C(lit.) |
Buffer component, may be used for the removal of peripheral membrane proteins.
1 kg in poly bottle
5 kg in poly drum
500 g in poly bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed

powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?