CAS Number 9005-25-8
Linear Formula (C6H10O5)n
EC Number 232-679-6
MDL number MFCD00082026
PubChem Substance ID 24899534
| Related Categories | Biochemicals and Reagents, Carbohydrate, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, |
Heat hydrolyzed
Customers Also Viewed


Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?