EMAIL THIS PAGE TO A FRIEND
S2804 Sigma-Aldrich

Sodium sulfate

meets USP testing specifications

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z, Pharmacopoeial Inorganics
grade   meets USP testing specifications
InChI Key   PMZURENOXWZQFD-UHFFFAOYSA-L
assay   99.5% dry basis (NLT)
form   crystalline
mp   884 °C(lit.)
solubility   water: soluble 185 g/L
density   2.68 g/mL at 25 °C(lit.)
suitability   meets USP testing specifications

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WE1650000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?