• USA Home
  • S2929 - Sodium thiosulfate pentahydrate

EMAIL THIS PAGE TO A FRIEND
S2929 Sigma-Aldrich

Sodium thiosulfate pentahydrate

meets USP testing specifications

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z, Pharmacopoeial Inorganics
InChI Key   PODWXQQNRWNDGD-UHFFFAOYSA-L
suitability   meets USP testing specifications

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WE6660000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?