Synonym: Nitric acid silver(I) salt
CAS Number 7761-88-8
Linear Formula AgNO3
Molecular Weight 169.87
EC Number 231-853-9
MDL number MFCD00003414
PubChem Substance ID 24899707
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Essential Chemicals, Hematology and Histology, Inorganic Salts, Solutions and Reagents, Reagent Plus, Research Essentials, |
| grade | ReagentPlus® |
| vapor density | 5.8 (vs air) |
| assay | ≥99.0% (titration) |
| form | solid |
| mp | 212 °C (dec.)(lit.) |
5, 25, 100 g in amber poly
500 g in poly bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?