EMAIL THIS PAGE TO A FRIEND
S9627 Sigma-Aldrich

Sodium sulfate

ReagentPlus®, ≥99.0%

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Salts
grade   ReagentPlus®
assay   ≥99.0%
form   powder
loss   ≤0.5% loss on drying
mp   884 °C(lit.)
solubility   water: soluble1 g/10 mL
density   2.68 g/mL at 25 °C(lit.)

Description

Packaging

2.5 kg in poly bottle

5, 10 kg in poly drum

500 g in poly bottle

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Other Notes

Go to Redi-Dri Product Listing 793531

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WE1650000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Determination of Water Content in Sodium sulfate, anhydrous Using Karl Fischer Titration

Adherent water in salts can be easily determined. It is best to choose a working medium which will prevent the salt from dissolving, e.g. a Methanol-Chloroform mixture with an excess of Chloroform. T...
Keywords: Crystallization, Titrations

Protocols

Enzymatic Assay of Neuraminidase

2.1. This procedure applies to products that have a specification for neuraminidase content by enzymatic determination.

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?