Synonym: Disodium hydrogen arsenate heptahydrate, di-Sodium hydrogen arsenate heptahydrate
CAS Number 10048-95-0
Linear Formula Na2HAsO4 · 7H2O
Molecular Weight 312.01
MDL number MFCD00149179
PubChem Substance ID 24899852
| Related Categories | ACS Grade, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Research Essentials |
| grade | ACS reagent |
| assay | 98.0-102.0% |
| impurities | ≤0.005% Insoluble matter |
| anion traces | arsenite (As2O3): ≤0.01% |
| chloride (Cl-): ≤0.001% | |
| nitrate (NO3-): ≤0.005% | |
| sulfate (SO42-): ≤0.01% |
50 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?