EMAIL THIS PAGE TO A FRIEND
T0695 Sigma-Aldrich

True Blue Chloride

Synonym: 2,2′-(1,2-ethenediyl)bis-dihydrochloride dihydrochloride, 5-Benzofurancarboximidamide

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes, T-U-V More...
InChI Key   WFZFMHDDZRBTFH-CZEFNJPISA-N

Description

Application

UV light-excitable, divalent cationic dye stains the cytoplasm with blue fluorescence. Shows stability in immunohistochemical processing.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?