• USA Home
  • T0899 - Stearyl trichloroacetate

EMAIL THIS PAGE TO A FRIEND
T0899 Sigma-Aldrich

Stearyl trichloroacetate

~95%

Synonym: C18, Trichloroacetic acid stearyl ester

  • CAS Number 35425-17-3

  • Empirical Formula (Hill Notation) C20H37Cl3O2

  • Molecular Weight 415.87

  •  MDL number MFCD00063323

  •  PubChem Substance ID 24899921

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
InChI Key   SHVKFBUBFSLWFV-UHFFFAOYSA-N
assay   ~95%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
density   1.03 g/mL at 25 °C(lit.)
format   neat
Gene Information   human ... CA1(759), CA2(760), F2(2147), PRSS1(5644)

Description

Application

For use as retention index standards in gas chromatography.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?