EMAIL THIS PAGE TO A FRIEND
T112 Sigma-Aldrich

Tracazolate

analytical standard

Synonym: 4-(Butylamino)-1-ethyl-6-methyl-1H-pyrazolo(3,4-b)pyridine-5-carboxylic acid ethyl ester

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Structure, Chromatography,
grade   analytical standard
InChI Key   PCTRYMLLRKWXGF-UHFFFAOYSA-N
form   neat
  solid
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   white
solubility   0.1 M HCl: soluble
  0.1 M NaOH: slightly soluble
  H2O: slightly soluble
  methanol: soluble
format   neat

Description

Biochem/physiol Actions

Non-benzodiazepine anxiolytic which has a high differential between sedative and anxiolytic doses.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?