• USA Home
  • T3053 - Trypsin TPCK treated from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
T3053 Sigma-Aldrich

Trypsin TPCK treated from bovine pancreas

Purchase

Properties

Quality Level   GMP
storage temp.   −20°C

Description

Analysis Note

Protein determined by E1%/280 unless otherwise indicated.

General description

The trypsin molecule has two domains: one is related to the enzyme active site and the tryptophan residues; the other is related to the 8-anilinonaphthalene-1-sulfonate binding.

Unit Definition

One BAEE unit will produce a ΔA253 of 0.001 per min at pH_7.6 at 25° C using BAEE as substrate. Reaction volume = 3.2_ml (1_cm light path).

Application

Trypsin has been used in a study to assess the role of the Agr-like quorum sensing system in regulating toxin production by Clostridium perfringens type B strains. Trypsin has also been used in a study to investigate the identification of protease inhibitors using a fast fluorimetric assay.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YN5075000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?