Synonym:
3,7-
CAS Number 78338-22-4
Empirical Formula (Hill Notation) C12H9N3S · C2H4O2
Molecular Weight 287.34
Beilstein Registry Number 4345073
Colour Index Number 52000
MDL number MFCD00081194
PubChem Substance ID 24900137
Popular Documents: Product Information Sheet (PDF)
| composition | Dye content, ~90% |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?